
1,5-Diphenylcarbazide ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:140-22-7
:
Formula:C₁₃H₁₄N₄O
MDL Number:MFCD00003013
:242.28 g/mol
Merck Index:14,3321
Boiling Pt:454 °C (1013 hPa)
Melting Pt:170…176 °C
Density:1.31 g/cm³ (20 °C)
EINECS:205-403-7
:GHS07
Storage Temperature:Ambient
Related Information
Certificates
Search by Lot NumberALFAA14060.22_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Frequently Bought Together