Order Entry
Czech Republic
ContactUsLinkComponent
Chroman-4-one, Sigma-Aldrich®
Chroman-4-one, Sigma-Aldrich®
Catalog # SIAL122351-10G
Supplier:  Merck
CAS Number:  
Chroman-4-one, Sigma-Aldrich®
Catalog # SIAL122351-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :148.16 g/mol
Boiling Pt:253 °C (1013 hPa)
Melting Pt:39 °C
Density:1.1291 g/cm³ (99.8 °C)
 :M-Clarity™ quality level = MQ100
EINECS:207-736-3
CAS Number:491-37-2
Formula:C₉H₈O₂
MDL Number:MFCD00006840