

o-Terphenyl for synthesis, Sigma-Aldrich®
: Merck
:84-15-1
:
:C₁₈H₁₄
:MFCD00003055
:230.31 g/mol
:338 °C (1013 hPa)
:54…56 °C
:1.1 g/cm³ (20 °C)
:163 °C
:2811
:6.1,III
:M-Clarity™ quality level = MQ200
:201-517-6
:GHS06
:GHS09
:Ambient
- Solubility:- 0.01 g/l (25 °C)
- Melting Point:55 - 57 °C
- Molar Mass:230.3 g/mol
- Boiling Point:332 °C (1013 hPa)
- Flash Point:163 °C
Frequently Bought Together