Order Entry
Czech Republic
ContactUsLinkComponent
2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208)
2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208)
Catalog # PROOCIL-PCB-208-C
CAS Number:  
2,2',3,3',4,5,5',6,6'-Nonachlorobiphenyl (PCB No. 208)
Catalog # PROOCIL-PCB-208-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :464.21 g/mol
CAS Number:52663-77-1
Formula:C₁₂HCl₉