Order Entry
Czech Republic
ContactUsLinkComponent
2,4,6-Trichlorobiphenyl (PCB No. 30)
2,4,6-Trichlorobiphenyl (PCB No. 30)
Catalog # EHERC20003000
Supplier:  EHRENSTORFER
CAS Number:  
2,4,6-Trichlorobiphenyl (PCB No. 30)
Catalog # EHERC20003000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :257.55 g/mol
UN:2315
ADR:9,II
CAS Number:35693-92-6
Formula:C₁₂H₇Cl₃