Order Entry
Czech Republic
ContactUsLinkComponent
2,3,4,6-Tetrachlorobiphenyl (PCB No. 62)
2,3,4,6-Tetrachlorobiphenyl (PCB No. 62)
Catalog # PROOCIL-PCB-62-C
CAS Number:  
2,3,4,6-Tetrachlorobiphenyl (PCB No. 62)
Catalog # PROOCIL-PCB-62-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
CAS Number:54230-22-7
Formula:C₁₂H₆Cl₄