3,4,4'-Trichlorobiphenyl (PCB No. 37)
Catalog # PROOCIL-PCB-37-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:257.55 g/mol
CAS Number:38444-90-5
Formula:C₁₂H₇Cl₃