Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4',6-Trichlorobiphenyl (PCB No. 32)
2,4',6-Trichlorobiphenyl (PCB No. 32)
2,4',6-Trichlorobiphenyl (PCB No. 32)
2,4',6-Trichlorobiphenyl (PCB No. 32)
Catalog # PROOCIL-PCB-32-C
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Expand image
2,4',6-Trichlorobiphenyl (PCB No. 32)
Catalog # PROOCIL-PCB-32-C
Supplier:
LGC Standards PROMOCHEM
CAS Number:
Specifications
Pk:
1
Seq:
0001
Specifications
About this item
Product Details
:
:
257.55 g/mol
CAS Number:
38444-77-8
Formula:
C₁₂H₇Cl₃
Product Details