Order Entry
Czech Republic
ContactUsLinkComponent
 
2,4,4'-Trichlorobiphenyl (PCB No. 28)
Catalog # PROOBCR-291
CAS Number:  
2,4,4'-Trichlorobiphenyl (PCB No. 28)
Catalog # PROOBCR-291
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:9,II
 :257.55 g/mol
Storage Temperature:Refrigerator
EINECS:230-293-2
UN:2315
CAS Number:7012-37-5
Formula:C₁₂H₇Cl₃