2,4,4',5-Tetrachlorobiphenyl (PCB No. 74)
Catalog # PROOCIL-PCB-74-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:291.99 g/mol
CAS Number:32690-93-0
Formula:C₁₂H₆Cl₄