Order Entry
Czech Republic
ContactUsLinkComponent
2,3,3'-Trichlorobiphenyl (PCB No. 20)
2,3,3'-Trichlorobiphenyl (PCB No. 20)
Catalog # PROOBCR-290
CAS Number:  
2,3,3'-Trichlorobiphenyl (PCB No. 20)
Catalog # PROOBCR-290
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:9,II
 :257.55 g/mol
UN:3432
CAS Number:38444-84-7
Formula:C₁₂H₇Cl₃