2,3,3',4,4',6-Hexachlorobiphenyl (PCB No. 158)
Catalog # PROOCIL-PCB-158-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
CAS Number:74472-42-7
Formula:C₁₂H₄Cl₆