Order Entry
Czech Republic
ContactUsLinkComponent
2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)
2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)
Catalog # EHERC20015600
Supplier:  EHRENSTORFER
CAS Number:  
2,3,3',4,4',5-Hexachlorobiphenyl (PCB No. 156)
Catalog # EHERC20015600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :360.88 g/mol
Storage Temperature:Refrigerator
UN:3432
ADR:9,II
CAS Number:38380-08-4
Formula:C₁₂H₄Cl₆