Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151)
2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151)
2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151)
PROOCIL-PCB-151-C
:
LGC Standards PROMOCHEM
:
2,2',3,5,5',6-Hexachlorobiphenyl (PCB No. 151)
PROOCIL-PCB-151-C
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
2210
:
:
360.88 g/mol
:
52663-63-5
:
C₁₂H₄Cl₆