2,3',4,4',5'-Pentachlorobiphenyl (PCB No. 123)
Catalog # EHERC20012300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:326.44 g/mol
CAS Number:65510-44-3
Formula:C₁₂H₅Cl₅