Order Entry
Czech Republic
ContactUsLinkComponent
 
2,2',4,4',5-Pentachlorobiphenyl (PCB No. 99)
Catalog # PROOCIL-PCB-99-C
CAS Number:  
2,2',4,4',5-Pentachlorobiphenyl (PCB No. 99)
Catalog # PROOCIL-PCB-99-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :326.44 g/mol
CAS Number:38380-01-7
Formula:C₁₂H₅Cl₅