Order Entry
Czech Republic
ContactUsLinkComponent
2,2',3,4,4',6-Hexachlorobiphenyl (PCB No. 139)
2,2',3,4,4',6-Hexachlorobiphenyl (PCB No. 139)
Catalog # PROOCIL-PCB-139-C
CAS Number:  
2,2',3,4,4',6-Hexachlorobiphenyl (PCB No. 139)
Catalog # PROOCIL-PCB-139-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
CAS Number:56030-56-9
Formula:C₁₂H₄Cl₆