2,2',3,3',5-Pentachlorobiphenyl (PCB No. 83)
Catalog # PROOCIL-PCB-83-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:326.44 g/mol
CAS Number:60145-20-2
Formula:C₁₂H₅Cl₅