Order Entry
Czech Republic
ContactUsLinkComponent
 
2,2',3,3',4-Pentachlorobiphenyl (PCB No. 82)
Catalog # EHERE20008200
Supplier:  EHRENSTORFER
CAS Number:  
2,2',3,3',4-Pentachlorobiphenyl (PCB No. 82)
Catalog # EHERE20008200
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :326.44 g/mol
CAS Number:52663-62-4
Formula:C₁₂H₅Cl₅