Order Entry
Czech Republic
ContactUsLinkComponent
3,5-Dichlorobiphenyl (PCB No. 14)
3,5-Dichlorobiphenyl (PCB No. 14)
Catalog # EHERC20001400
Supplier:  EHRENSTORFER
CAS Number:  
3,5-Dichlorobiphenyl (PCB No. 14)
Catalog # EHERC20001400
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :223.1 g/mol
UN:3432
ADR:9,II
CAS Number:34883-41-5
Formula:C₁₂H₈Cl₂
MDL Number:MFCD00055256