Order Entry
Czech Republic
ContactUsLinkComponent
3,3',5,5'-Tetrachlorobiphenyl (PCB No. 80)
3,3',5,5'-Tetrachlorobiphenyl (PCB No. 80)
Catalog # PROOCIL-PCB-80-C
CAS Number:  
3,3',5,5'-Tetrachlorobiphenyl (PCB No. 80)
Catalog # PROOCIL-PCB-80-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
CAS Number:33284-52-5
Formula:C₁₂H₆Cl₄