Order Entry
Czech Republic
ContactUsLinkComponent
2,2',4,6'-Tetrachlorobiphenyl (PCB No. 51)
2,2',4,6'-Tetrachlorobiphenyl (PCB No. 51)
Catalog # PROOCIL-PCB-51-C
CAS Number:  
2,2',4,6'-Tetrachlorobiphenyl (PCB No. 51)
Catalog # PROOCIL-PCB-51-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
CAS Number:68194-04-7
Formula:C₁₂H₆Cl₄