Order Entry
Czech Republic
ContactUsLinkComponent
2,6-Dichlorobiphenyl (PCB No. 10)
2,6-Dichlorobiphenyl (PCB No. 10)
Catalog # PROOCIL-PCB-10-C
CAS Number:  
2,6-Dichlorobiphenyl (PCB No. 10)
Catalog # PROOCIL-PCB-10-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
ADR:9,II
 :223.1 g/mol
UN:3432
CAS Number:33146-45-1
Formula:C₁₂H₈Cl₂