3,3',4,4'-Tetrachlorobiphenyl (PCB No. 77)
Catalog # EHERC20007700
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:291.99 g/mol
Boiling Pt:381 °C (1013 hPa)
Melting Pt:173 °C
UN:3432
ADR:9,II
CAS Number:32598-13-3
Formula:C₁₂H₆Cl₄