Order Entry
Czech Republic
ContactUsLinkComponent
2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56)
2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56)
Catalog # EHERC20005600
Supplier:  EHRENSTORFER
CAS Number:  
2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56)
Catalog # EHERC20005600
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :291.99 g/mol
CAS Number:41464-43-1
Formula:C₁₂H₆Cl₄