Order Entry
Czech Republic
ContactUsLinkComponent
2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56)
2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56)
Catalog # PROOCIL-PCB-56-C
CAS Number:  
2,3,3',4'-Tetrachlorobiphenyl (PCB No. 56)
Catalog # PROOCIL-PCB-56-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :291.99 g/mol
CAS Number:41464-43-1
Formula:C₁₂H₆Cl₄