Order Entry
Czech Republic
ContactUsLinkComponent
 
2,2',3,3'-Tetrachlorobiphenyl (PCB No. 40)
Catalog # EHERC20004000
Supplier:  EHRENSTORFER
CAS Number:  
2,2',3,3'-Tetrachlorobiphenyl (PCB No. 40)
Catalog # EHERC20004000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :291.99 g/mol
UN:3432
ADR:9,II
CAS Number:38444-93-8
Formula:C₁₂H₆Cl₄