2,3,3',4',5,5'-Hexachlorobiphenyl (PCB No. 162)
Katalogové číslo PROOCIL-PCB-162-C
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:360.88 g/mol
CAS Number:39635-34-2
Formula:C₁₂H₄Cl₆