Order Entry
Czech Republic
ContactUsLinkComponent
2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133)
2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133)
Catalog # PROOCIL-PCB-133-C
CAS Number:  
2,2',3,3',5,5'-Hexachlorobiphenyl (PCB No. 133)
Catalog # PROOCIL-PCB-133-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
CAS Number:35694-04-3
Formula:C₁₂H₄Cl₆