Order Entry
Czech Republic
ContactUsLinkComponent
2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167)
2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167)
Catalog # PROOCIL-PCB-167-C
CAS Number:  
2,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 167)
Catalog # PROOCIL-PCB-167-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
Boiling Pt:383 °C (1013 hPa)
Melting Pt:125…127 °C
UN:3432
ADR:9,II
CAS Number:52663-72-6
Formula:C₁₂H₄Cl₆