Order Entry
Czech Republic
ContactUsLinkComponent
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
Catalog # PROOBCR-297
CAS Number:  
2,2',4,4',5,5'-Hexachlorobiphenyl (PCB No. 153)
Catalog # PROOBCR-297
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :360.88 g/mol
Boiling Pt:259 °C (1013 hPa)
Melting Pt:103…104 °C
Storage Temperature:Refrigerator
UN:3432
ADR:9,II
CAS Number:35065-27-1
Formula:C₁₂H₄Cl₆