Order Entry
Czech Republic
ContactUsLinkComponent
3,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 169)
3,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 169)
Catalog # PROOCIL-PCB-169-C
CAS Number:  
3,3',4,4',5,5'-Hexachlorobiphenyl (PCB No. 169)
Catalog # PROOCIL-PCB-169-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
Boiling Pt:383 °C (1013 hPa)
Melting Pt:208…210 °C
CAS Number:32774-16-6
Formula:C₁₂H₄Cl₆