Order Entry
Czech Republic
ContactUsLinkComponent
 
2,2',3,3',4,6'-Hexachlorobiphenyl (PCB No. 132)
Catalog # PROOCIL-PCB-132-C
CAS Number:  
2,2',3,3',4,6'-Hexachlorobiphenyl (PCB No. 132)
Catalog # PROOCIL-PCB-132-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
CAS Number:38380-05-1
Formula:C₁₂H₄Cl₆