Zadání objednávky
Czech Republic
ContactUsLinkComponent
2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194)
2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194)
Catalog # PROOCIL-PCB-194-C
CAS Number:  
2,2',3,3',4,4',5,5'-Octachlorobiphenyl (PCB No. 194)
Catalog # PROOCIL-PCB-194-C
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle for solids
  • Bal.:
    5 mg
  • Sekvence:
    2210

Specifications

About this item

 :
UN:2315
ADR:9,II
CAS Number:35694-08-7
Formula:C₁₂H₂Cl₈