Order Entry
Czech Republic
ContactUsLinkComponent
2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166)
2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166)
Catalog # PROOCIL-PCB-166-C
CAS Number:  
2,3,4,4',5,6-Hexachlorobiphenyl (PCB No. 166)
Catalog # PROOCIL-PCB-166-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :360.88 g/mol
CAS Number:41411-63-6
Formula:(C₆H₄Cl)(C₆Cl₅)