0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
2,2',3,3',4,5,6-Heptachlorobiphenyl (PCB No. 173)
2,2',3,3',4,5,6-Heptachlorobiphenyl (PCB No. 173)
2,2',3,3',4,5,6-Heptachlorobiphenyl (PCB No. 173)
PROOCIL-PCB-173-C
:
LGC Standards PROMOCHEM
:
2,2',3,3',4,5,6-Heptachlorobiphenyl (PCB No. 173)
PROOCIL-PCB-173-C
:
LGC Standards PROMOCHEM
:
Pack type:
Glass bottle for solids
Pk:
5 mg
Seq:
2210
:
:
395.32 g/mol
:
68194-16-1
:
(C₆H₃Cl₂)C₆Cl₅