
2,4,6-Triisopropylbenzenesulphonyl chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:6553-96-4
:
Formula:C₁₅H₂₃ClO₂S
MDL Number:MFCD00007433
:302.87 g/mol
Melting Pt:94…97 °C
Storage Temperature:Ambient
EINECS:229-479-6
UN:3261
ADR:8,II
:GHS05
Related Information
Frequently Bought Together