2',5'-Dimethylacetophenone, Sigma-Aldrich®
Catalog # SIAL454265-5ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Seq:2210
Specifications
About this item
:
Boiling Pt:231 °C (1013 hPa)
Melting Pt:–18 °C
Density:1.0154 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ100
:148.2 g/mol
EINECS:218-399-7
CAS Number:2142-73-6
Formula:C₁₀H₁₂O
MDL Number:MFCD00014988