Order Entry
Czech Republic
ContactUsLinkComponent
4,4'-Bis(dimethylamino)benzophenone for synthesis, Sigma-Aldrich®
Corrosive
Health
4,4'-Bis(dimethylamino)benzophenone for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Corrosive
Health
4,4'-Bis(dimethylamino)benzophenone for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    0.4 g/l (20 °C)
  • Melting Point:
    172 - 175 °C
  • Molar Mass:
    268.35 g/mol
  • Bulk Density:
    400 kg/m3
  • Boiling Point:
    360 °C (1013 hPa)
  • Flash Point:
    220 °C
  • Ignition Point:
    480 °C

Specification Test Results

Related Information

Frequently Bought Together