2,5-Di-tert-butylhydroquinone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:88-58-4
:
Formula:C₁₄H₂₂O₂
MDL Number:MFCD00008825
:222.33 g/mol
Boiling Pt:334 °C (1013 hPa)
Melting Pt:212 °C
Density:1.078 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-841-8
Frequently Bought Together