
2,6-Di-tert-butyl-4-methoxyphenol 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:489-01-0
:
Formula:C₁₅H₂₄O₂
MDL Number:MFCD00008824
:236.35 g/mol
Boiling Pt:135…140 °C (10 mmHg)
Melting Pt:103…106 °C
EINECS:207-693-0
:GHS07
Specification Test Results
- Appearance:White to beige Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:102 to 106 °C
- GC:≥96.0 %
Specification Test Results
Frequently Bought Together