ContactUsLinkComponent
J.T.Baker® BAKERBOND spe™ Cyano (CN), SPE Columns
J.T.Baker® BAKERBOND spe™ Cyano (CN), SPE Columns
Supplier:  Avantor
Certificates
undefined
J.T.Baker® BAKERBOND spe™ Cyano (CN), SPE Columns
Supplier:  Avantor
Certificates
Specifications
  • Catalog No:
  • 7021-07
  • 7021-02
  • Carbon load:
  • 10,5%
  • 10,5%
  • Endcapped:
  • Yes
  • Yes