3,5-Bis(trifluoromethyl)phenyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:23165-29-9
:
Formula:C₉H₃F₆NS
MDL Number:MFCD00040838
:271.19 g/mol
Boiling Pt:94…95 °C (19 mmHg)
Density:1.48 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Frequently Bought Together