

4'-(Trifluoromethyl)acetophenone 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:709-63-7
:
Formula:C₉H₇F₃O
MDL Number:MFCD00000401
:188.15 g/mol
Boiling Pt:79…80 °C (8 mmHg)
Melting Pt:30…33 °C
Density:0.92 g/cm³
Flash Pt:84 °C
Storage Temperature:Ambient
EINECS:211-913-0
UN:1325
ADR:4.1,II
:GHS07
:GHS02
Specification Test Results
- Appearance:White to light yellow Low melting solid
- Infrared spectrum:Conforms
- Melting point:30 to 33 °C
- GC:≥98.5 %
Specification Test Results
Frequently Bought Together