2'-(Trifluoromethyl)acetophenone
Catalog # ABCRAB104277.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:188.15 g/mol
Boiling Pt:163 °C (1013 hPa)
Melting Pt:15…17 °C
Density:1,25 g/cm³ (20 °C)
EINECS:241-434-2
CAS Number:17408-14-9
Formula:C₉H₇F₃O
MDL Number:MFCD00000378