
4'-Methylacetophenone for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:122-00-9
:
Formula:C₉H₁₀O
MDL Number:MFCD00008751
:134.18 g/mol
Boiling Pt:226 °C (1013 hPa)
Melting Pt:28 °C
Density:1.013 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:92 °C
EINECS:204-514-8
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:2.07 g/l (20 °C)
- Melting Point:28 °C
- Molar Mass:134.18 g/mol
- Boiling Point:226 °C (1013 hPa)
- Vapor Pressure:0.14 hPa (20 °C)
- Flash Point:92 °C
- Refractive Index:1.5313 (25 °C, 589 nm)
- Explosion Limit:1.00 %(V)
- Density:1.00 g/cm3 (20 °C)
- Synonyms:Methyl p-tolyl ketone
- Hill Formula:C9H10O
Specification Test Results
Frequently Bought Together