2'-Methylacetophenone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:577-16-2
:
Formula:C₉H₁₀O
MDL Number:MFCD00008734
:134.18 g/mol
Boiling Pt:214 °C (1013 hPa)
Density:1.0262 g/cm³ (0 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:209-408-5
Frequently Bought Together