
4-Bromo-2,6-dimethylanisole ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14804-38-7
:
Formula:C₉H₁₁BrO
MDL Number:MFCD00143259
:215.09 g/mol
Boiling Pt:117…120 °C (13 mmHg)
Density:1.347
Flash Pt:>110 °C (230 °F)
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together