
2-Methylbiphenyl, Sigma-Aldrich®
Catalog # SIAL292192-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:168.24 g/mol
Boiling Pt:255 °C (1013 hPa)
Melting Pt:–0.2 °C
Density:1.011 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:211-400-1
:GHS07
CAS Number:643-58-3
Formula:C₁₃H₁₂
MDL Number:MFCD00008517