4-Biphenylylmethanol, Sigma-Aldrich®
Catalog # SIAL123838-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:184.24 g/mol
Boiling Pt:183…184 °C (11 mmHg)
Melting Pt:104…105 °C
:M-Clarity™ quality level = MQ100
EINECS:222-745-2
CAS Number:3597-91-9
Formula:C₁₃H₁₂O
MDL Number:MFCD00004660